Imprimer
Homogeniser, FastPrep®-96, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
116010560 116010570 116010550 116010500
ICNA116010560EA 1998 EUR
ICNA116010560 ICNA116010570 ICNA116010550 432-0365
Homogeniser, FastPrep®-96, MP Biomedicals
Homogénéisateurs Homogénéisateurs de paillasse
FastPrep-96™ is a versatile high speed homogeniser offering the ultimate in high throughput sample preparation. It is used to homogenise biological samples. FastPrep®-96 lyses thoroughly and quickly any tissues and cells so allowing easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
  • High throughput process up to 192 samples simultaneously in 2×96 deep well plates
  • Exceptional versatility interchangeable adapters available for 2×96 deep well plates, 96×2 ml tubes, 48×4,5 ml tubes, 24×15 ml tubes, 8×50 ml tubes, and 1×250 ml flask
  • Excellent reproducibility consistent lysis results in every well with automated feedback control
  • Fast processing speed 1800 oscillations/min and 1,5-inch stroke - highest available
  • True Linear Motion Eliminates the need to re-orient plates mid-cycle Preservation of temperature sensitive molecules, homogenisation performed at room or cryogenic temperatures
Developed for resistant samples, the FastPrep-96™ instrument uses a high speed linear motion to thoroughly disrupt any tissues and cells in just seconds through the simultaneous beating of specialised Lysing Matrix beads on the sample material. This high performance system saves hours of work during the sample preparation stage and allows for the purification of high yields of intact DNA, RNA, proteins and other metabolites.