Imprimer
Ion selective electrodes, cyanide, Orion™
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9406BN 9606BNWP
662-0825EA 1100 EUR
662-0825 662-1593
Ion selective electrodes, cyanide, Orion™
Électrodes Électrodes ISE
These electrodes are designed for measuring cyanide ions in aqueous solutions quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes
Informations de commande: The half cell electrodes require a separate reference electrode which can be ordered separately.