Imprimer
Pure quartz air sampling filters, grade QM-H, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1853-047-50 1853-150-50 1853-090-50 1853-037-50 1853-050-50
513-0094EA 272 EUR
513-0094 513-0125 513-0124 513-0122 513-0123
Pure quartz air sampling filters, grade QM-H, Whatman™
Filtres Filtres de fibres de verre
Whatman™ QM-H is a pure quartz fibre filter for use in testing acidic gases, stacks, flues and aerosols. It has low heavy metal content so it is well suited for heavy metals analysis.
  • High stability up to 900 °C
  • High filtration efficiency
  • Binder-free
  • Minimal reactivity with biological or chemical components
Pure quartz fiber filters come in 50-piece packs with five diameters: 37 mm, 47 mm, 50 mm, 90 mm, 150 mm.
Commander maintenant
SPÉCIFICATIONS
QM-H
Environmentally Preferable
Poids 85 g/m²
Épaisseur 430 µm
Max. temperature 900 °C