AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Marques : MP BiomedicalsTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AlbumiNZ™ protease-reduced BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality.
- Off-white to light yellowish to brown powder
- With ≤5% of moisture content
- Dissolves in ≤20 min at 20 to 25 °C (20% w/w in water)
AlbumiNZ™ protease-reduced BSA is useful for immunoassays where proteases may degrade proteins or where peroxidase or alkaline phospahatase may hydrolyse substrates.