Imprimer
Ion selective electrodes, potassium, Orion™
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9319BN 9719BNWP
662-0817EA 933 EUR
662-0817 662-1578
Ion selective electrodes, potassium, Orion™
Électrodes Électrodes ISE
These potassium electrodes are designed to measure potassium in aqueous solutions simply, accurately and economically.
  • Built-in Sure-Flow™ reference junction prevents electrode clogging and provides fast and stabile readings
  • Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes
Informations de commande: The half cell electrode requires a separate reference electrode which can be ordered separately.