Imprimer
Cleaning trolley, Clino® STEC 20, with MultiBox and PlusBox
Fournisseur: PFENNIG REINIGUNGSTECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3202000 3205000
PPSA3202000EA 909 EUR
PPSA3202000 PPSA3205000
Cleaning trolley, Clino® STEC 20, with MultiBox and PlusBox
Trolleys
The Clino® STEC20 is a compact system trolley made of stainless steel used for offices, shops and buldings with less space requirements.
  • Alternating between one and two-stage process is possible
  • Floor cleaning is simple with flat wringer Ringo and mop-reuse-technique
  • MultiBox with enormous storage space, extendable and lockable
  • PluBox system for the easy impregnation of mop covers and cloth (suitable for 6 L boxes)
  • Disposal unit at rest can be folded in, every time exchangeable and expandable
  • Double ball-bearinged guide wheels (125 mm)
MultiBox with its enclosed appearance and open function are used for the simple pre-preparation of mop heads and cloths. Both MultiBox and PlusBox are available in different colour.
Informations de livraison: Supplied with 1×Clino MultiBox, bottle holder ClinoBar, disposal MultiBoxes and disposal disconnectable device. PluBox includes with a sieve and cover for higher safety in manual cleaning and a straight preparation of all textiles.