Imprimer
Lave-verres de laboratoire sous paillasse, PG 8593
Fournisseur: MIELE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10085390 10085400 10084340 10085460 10087410 10088820 9960910 10084520 10114050 10114010 10124700 10096870 10096900 10114330 10138400 10138410 10138420 10114140 10114150 10114200 10114260 10411700 10782220
141-1284EA 0 EUR
141-1284 MIEL10085400 141-1288 MIEL10085460 141-1279 141-1278 141-1299 141-1239 141-1248 141-1245 MIEL10124700 141-1254 141-1253 MIEL10114330 MIEL10138400 MIEL10138410 MIEL10138420 141-1303 MIEL10114150 141-1385 141-1388 141-1450 141-1451
Lave-verres de laboratoire sous paillasse, PG 8593
Lave-verres
Miele PG 8593 laboratory glasswasher represents a professional solution for laboratory glassware for analytical experiments. This approach is particularly gentle on materials and is recommended by leading manufacturers of laboratory glassware.
  • Touch on metal' controls with three-line text display such as program name, actual/target temperature and remaining program time
  • Program catering for all disciplines and vacant program slots for user-specific programs
  • Freely assignable direct access buttons and individually selectable program names for fast access to
En savoir plus
Commander maintenant
SPÉCIFICATIONS
Modèle PG 8593 PG 8593 PG 8593 PG 8593 PG 8593 PG 8593
Environmentally Preferable
Capacité 128 flacons à col étroit ou 98 pipettes 128 flacons à col étroit ou 98 pipettes 128 flacons à col étroit ou 98 pipettes 128 flacons à col étroit ou 98 pipettes 128 flacons à col étroit ou 98 pipettes 128 flacons à col étroit ou 98 pipettes
Commande/programmes Panneau avec écran TouchControl / 15 programmes (2 positions programmables à volonté)
Mécanismes de nettoyage Deux bras d’aspersion (un troisième dans le panier supérieur)
Doseurs 1 pompe de dosage pour les agents nettoyants liquides est intégrée 1 pompe de dosage pour les agents nettoyants liquides est intégrée
Pompe de circulation 500 L/min 500 L/min 500 L/min 500 L/min 500 L/min 500 L/min
Distributeurs 1 dispenser pump for neutralising agent;
1 connection for DOS K 85 or DOS K 85/1 liquid dispensers (DOS must be ordered)
1 dispenser pump for neutralising agent;
1 connection for DOS K 85 or DOS K 85/1 liquid dispensers (DOS must be ordered)
1 dispenser pump for neutralising agent;
1 connection for DOS K 85 or DOS K 85/1 liquid dispensers (DOS must be ordered)
1 dispenser pump for neutralising agent;
1 connection for DOS K 85 or DOS K 85/1 liquid dispensers (DOS must be ordered)
1 dispenser pump for neutralising agent;
1 connection for DOS K 85 or DOS K 85/1 liquid dispensers (DOS must be ordered)
1 dispenser pump for neutralising agent;
1 connection for DOS K 85 or DOS K 85/1 liquid dispensers (DOS must be ordered)
Exigence électrique 3 N, 400 V AC, 50 Hz;
Fuse: 3×16 A;
Convertible to: 1 N, 230 V AC, 50 Hz
3 N, 400 V AC, 50 Hz;
Fuse: 3×16 A;
Convertible to: 1 N, 230 V AC, 50 Hz
3 N, 400 V AC, 50 Hz;
Fuse: 3×16 A;
Convertible to: 1 N, 230 V AC, 50 Hz
3 N, 400 V AC, 50 Hz;
Fuse: 3×16 A;
Convertible to: 1 N, 230 V AC, 50 Hz
3 N, 400 V AC, 50 Hz;
Fuse: 3×16 A;
Convertible to: 1 N, 230 V AC, 50 Hz
3 N, 400 V AC, 50 Hz;
Fuse: 3×16 A;
Convertible to: 1 N, 230 V AC, 50 Hz
Chauffage 8,5 kW 8,5 kW 8,5 kW 8,5 kW 8,5 kW 8,5 kW
Plomberie 1× cold water (CW);
1× cold water for steam condenser;
1× de-mineralised water (AD);
1× hot water (HW);
1 drain hose connection
1× cold water (CW);
1× cold water for steam condenser;
1× de-mineralised water (AD);
1× hot water (HW);
1 drain hose connection
1× cold water (CW);
1× cold water for steam condenser;
1× de-mineralised water (AD);
1× hot water (HW);
1 drain hose connection
1× cold water (CW);
1× cold water for steam condenser;
1× de-mineralised water (AD);
1× hot water (HW);
1 drain hose connection
1× cold water (CW);
1× cold water for steam condenser;
1× de-mineralised water (AD);
1× hot water (HW);
1 drain hose connection
1× cold water (CW);
1× cold water for steam condenser;
1× de-mineralised water (AD);
1× hot water (HW);
1 drain hose connection
Alimentation Tension d’entrée 3N CA 400 V, 50 Hz, branchement électrique complet: 9,3 kW, Protection [A]: 3 x 16 ou CA 230 V, 50 Hz, branchement électrique complet: 3,30 kW, Protection [A]: 16
Charge nominale 6,2 kW 6,2 kW 6,2 kW 6,2 kW 6,2 kW 6,2 kW
Raccordements eau 1× eau froide (EC), 1× eau froide pour condensateur de vapeur, 1× eau déminéralisée (AD), 1× eau chaude (AW) 1× eau froide (EC), 1× eau froide pour condensateur de vapeur, 1× eau déminéralisée (AD), 1× eau chaude (AW) 1×eau froide (EC), 1×eau froide pour condensateur de vapeur, 1×eau déminéralisée (AD), 1×eau chaude (AW) 1× eau froide (EC), 1× eau froide pour condensateur de vapeur, 1× eau déminéralisée (AD), 1× eau chaude (AW)
L×P×H 600×600×835 mm 600×600×835 mm 600×600×835 mm 600×600×835 mm 600×600×835 mm 600×600×835 mm