Imprimer
Mineral Mixes, AIN-93G, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0296040002 0296040010 0296040050
ICNA0296040002EA 87.9 EUR
ICNA0296040002 ICNA0296040010 ICNA0296040050
Mineral Mixes, AIN-93G, MP Biomedicals
Fournitures pour animaux Alimentation animale
This mineral mix, part of AIN-93G rodent diet, is used for growth, pregnancy and lactational phases of rodents.
For complete diet composition, please see the product datasheet.