Imprimer
n-Dodécane ≥99%, liquide clair et incolore
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Dodecane
0219515880 0219515890 0219515891
ICNA0219515880EA 35.6 EUR
ICNA0219515880 ICNA0219515890 ICNA0219515891
n-Dodécane ≥99%, liquide clair et incolore
n-Dodécane
N-Dodecane is a liquid alkane hydrocarbon.
N-Dodecane is used as a solvent, distillation chaser, scintillator component. Moreover it is used as a diluent for tributyl phosphate (TBP) in plants reprocessing.
  • Presentation: Clear, Colorless Liquid
  • Auto Ignition: 203 °C (397 °F) (lit.)
  • Vapor Density: 5.96 (vs air) (Lit.)
  • Vapor Pressure: 1 mmHg ( 47.8 °C) (Lit.)
  • Refractive Index: n20/D 1.421(lit.)
  • Storage temperature: Store at Room Temperature (15-30 °C). Store Desiccated.
Formule: H₃C(CH₂)₁₀CH₃
Point d'ébullition: 215…217 °C (lit.)                      
Point de fusion: −9,6 °C (lit.)                      
Densité: 0,753 (25 °C) (lit.)                      
Température de stockage: Température ambiante
Numéro MDL: MFCD00008969
Numéro CAS: 112-40-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616d3d75a63f52fbe2a8be1681d9233be6dc0928&vid=3268fe23453ad703df2782bd3c7ba6b939ba00cc&prid=web&clid=SAPDAM
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard