Imprimer
Ficoll 400, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0216000305 0216000310 0216000325
ICNA0216000305EA 56.5 EUR
ICNA0216000305 ICNA0216000310 ICNA0216000325
Ficoll 400, MP Biomedicals
Milieux de séparation cellulaire
Ficoll® is a synthetic, hydrophilic polymer made by the copolymerisation of sucrose and epichlorhydrin. It is a highly branched polymer. Solutions at neutral pH can be sterilised by autoclaving.
  • MW: 400000 g/mol
FICOLL® has the follwing key advantages: Proven medium for density gradient centrifugation. It does not penetrate biological membranes and has high density solution of low viscosity and low osmotic pressure. Ir preserves functional and morphological integrity. It is useful for separating cells which are sensitive to centrifugation and for separating cells of similar density but different size (under conditions of sedimentation at unit gravity). It can be used in other applications such as electrophoresis, hybridisation cryopreservation and as a hapten carrier.