Imprimer
n-Undécane ≥99%, liquide clair et incolore
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Undecane
0215823180 0215823190
ICNA0215823180EA 63.5 EUR
ICNA0215823180 ICNA0215823190
n-Undécane ≥99%, liquide clair et incolore
n-Undécane
Undecane may be used as an internal standard in gas chromatography when working with other hydrocarbons. Since the boiling point of undecane (196 °C) is well known, it may be used as a comparison for retention times in a gas chromatograph for molecules whose structure has been freshly elucidated.
  • Japan fire code: 4.2
Formule: H₃C(CH₂)₉CH₃
Poids moléculaire: 156.31 g/mol
Point d'ébullition: 196
Point de fusion: -26.7
Point éclair: 60
Numéro MDL: MFCD00008959
Numéro CAS: 1120-21-4
UN: 2330
ADR: 3 III