Imprimer
(R)-(+)-Limonène, liquide
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): D(+)-Limonène, (R)-(+)-p-Mentha-1,8-diène, (R)-(+)-Dipentène, (R)-(+)-1-Méthyl-4-(1-méthylvinyl)cyclohexène
0215523480 0215523490
ICNA0215523480EA 47.5 EUR
ICNA0215523480 ICNA0215523490
(R)-(+)-Limonène, liquide
(R)-(+)-Limonène
D-Limonene is a chiral intermediate for natural product synthesis. It is used as a renewably based solvent in cleaning products.
D-Limonene is a monoterpene found to prevent mammary cancer by inducing hepatic glutathione-S- transferase and uridine diphosphoglucuronosyl transferase. D-Limonene is an isoprenylation inhibitor of small G-proteins.
Limonene is a cyclie terpene from Chinese medicinal herb essential oils used in the synthesis of carvone. Limonene may be used as a shrinking agent to dissolve polystyrene. Limonene may be used in various insecticidal and insect repellant applications.
Miscible with alcohol, with most fixed oils, mineral oil; very slightly soluble in glycerin; practically insoluble in water and in propyiene glycol.
Formule: C₁₀H₁₆
Densité: 0,84 g/ml
Température de stockage: Température ambiante
Numéro MDL: MFCD00062991
Numéro CAS: 5989-27-5
UN: 2052
ADR: 3,III