9(S)-HODE ≥98%
Marques : Cayman ChemicalTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
9-hydroxy-10,12-octadecadienoic acid, 9-hydroxylinoleic acid, 9-hydroxy-10,12-octadecadienoic acid, (E,E)-isomer
9(S)-HODE is produced by the lipoxygenation of linoleic acid in both plants and animals. It has been detected in atherosclerotic plaques, as an esterified component of membrane phospholipids and in oxidised LDL particles.
9(S)-HODE is supplied as a solution in ethanol. To change the solvent, simply evaporate the ethanol under a gentle stream of nitrogen and immediately add the solvent of choice. Solvents such as DMSO and dimethyl formamide purged with an inert gas can be used. The solubility of 9(S)-HODE in these solvents is approximately 50 mg/ml.
An organic solvent-free solution of 9(S)-HODE can be prepared by evaporating the ethanol and directly dissolving the neat oil in aqueous buffers. The solubility of 9(S)-HODE in PBS (pH 7,2) is approximately 1 mg/ml. Not recommend to store the aqueous solution for more than one day.
Formule:
C₁₈H₃₂O₃ |
Numéro MDL:
MFCD00065831 Numéro CAS: 73543-67-6 |
Résultats des tests de spécification
Purity | ≥98% |
Stability | ≥1 year at −20 °C |
Formulation | A Solution in Ethanol |
UV/Vis | λmax: 234 nm ε: 23000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...