Imprimer
Betamax™-ES liquid scintillation cocktail, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0188002001 0188002004
ICNA0188002001EA 303 EUR
ICNA0188002001 ICNA0188002004
Betamax™-ES liquid scintillation cocktail, MP Biomedicals
Cocktails de scintillation
BetaMax™ ES, as its name implies, is the most efficient environmentally safe LSC cocktail for beta particle counting in non-aqueous samples.
  • Non-toxic, Non-flammable and odorless
  • High tritium efficiency
  • Excellent with dry filters and test wipes