Imprimer
Cholesterol oxidase, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0215067425 0215067480 0215067483
ICNA0215067425EA 74.9 EUR
ICNA0215067425 ICNA0215067480 ICNA0215067483
Cholesterol oxidase, MP Biomedicals
Enzymes
CHOD is soluble in cold 50 mM potassium phosphate buffer, pH 7,0. Prepare solutions immediately before use.
One unit causes the formation of one µmole of hydrogen peroxide per minute at pH 7,0 and 37 °C.
Stabilisers: Bovine serum albumin, sugars. Typical Contaminants: Catalase: Not more than 1,0% Cholesterol esterase: Not more than 1,0 to 10²%. Michaelis Constant: 4,3×10⁻⁵ M (cholesterol). Inhibitors: Ionic detergents, Hg²⁺, Ag⁺; Optimum Temperature: 45 to 50 °C; thermal stability: below 45 °C (pH 7,0; 15 minutes).