Imprimer

Tétracycline Streptomyces species, powder_yellow

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

0210301205 0210301225 0210301280
ICNA0210301205EA 45 EUR
ICNA0210301205 ICNA0210301225 ICNA0210301280
Tétracycline Streptomyces species, powder_yellow
Tétracycline

Tetracyline is an antibiotic substance (gram-positive, gram-negative and mycoplasma) produced by streptomyces spp.


Tetracyline inhibits gram-positive, gram-negative and mycoplasma. It is used in tetracycline controlled gene expression systems (gene switches) such as the tet-on and tet-off systems. It is also used to select tetracyline resistant colonies in cell culture.


Tetracycline binds to the 30S subunit of the ribosome at the A-site. During protein biosynthesis, the new t-RNA with the amino acid attempts to bind to the A-site of the ribosome. However, since the A-site is blocked by the tetracycline, the aminoacyl-tRNA cannot bind to it. Without the attachment of the tRNA, protein biosynthesis cannot occur. Protein biosynthesis is inhibited and cell death occurs .


Soluble in water (1,7 mg/ml), or methanol (> 20 mg/ml - clear, yellow solution).


Attention: Research Use Only (RUO).

Formule: C₂₂H₂₄N₂O₈
Poids moléculaire: 444.43 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00151232
Numéro CAS: 60-54-8

Commander maintenant

Résultats des tests de spécification

Identity Test Passes
Potency ≥900 μg/mg
FTIR Conforms to Standard
UV/VIS: max (0.1N HCl) 215 - 220, 266 - 271, 354 - 357 nm
Expiration Dat Month/Day/Yea


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR