Imprimer
Lipase, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0210218980 0210218990
ICNA0210218980EA 57.2 EUR
ICNA0210218980 ICNA0210218990
Lipase, MP Biomedicals
Enzymes
One USP unit of lipase activity is contained in the amount of pancreatin that liberates 1,0 µEq of acid per minute at a pH of 9,0 at 37 °C under the conditions of the assay for lipase activity.
  • Partly soluble in water and insoluble in alcohol
Lipases are a subclass of the esterases. It catalyses the hydrolysis of emulsified esters of glycerol and long chain fatty acids.