Bovine Ribonuclease A (from Pancreas), MP Biomedicals
Marques : MP BiomedicalsTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Dissolves readily at 2 mg/ml in analytical grade water to give a clear colorless solution.
That amount of enzyme causing the hydrolysis of RNA at a rate such that k (velocity constant) equals unity at 25 °C and pH 5,0.