Imprimer
Mercaptamine chlorhydrate ∼98%, poudre blanche
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): 2-MEA
0210099505 0210099525 0210099580
ICNA0210099505EA 35.4 EUR
ICNA0210099505 ICNA0210099525 ICNA0210099580
Mercaptamine chlorhydrate ∼98%, poudre blanche
Mercaptamine chlorhydrate
β-Mercaptoethylamine (Cysteamine hydrochloride) is useful antioxidant. It is experimentally used as a radioprotective agent. It is also used to produce acute and chronic duodenal ulcers in rats and as an antidote to acetaminophen.
Soluble in water (50 mg/ml) clear, colorless to faint yellow, soluble in alcohol.
Attention: Research Use Only (RUO).
Formule: C₂H₇NS·HCl
Poids moléculaire: 113.61 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00012904
Numéro CAS: 156-57-0
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
Purity ~98%
FTIR Conforms to Standard
Melting Point 58 - 73 °C