Imprimer
Potassium hexacyanoferrate (II) trihydraté 98,5-102,0, powder_yellow crystalline ACS
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Potassium hexacyanoferrate (II) à 3 molécules d'eau, Potassium ferrocyanure trihydraté, Potassium cyanure jaune, Hexacyanoferrate de tétrapotassium trihydraté
0215256080 0215256090
ICNA0215256080EA 39.4 EUR
ICNA0215256080 ICNA0215256090
Potassium hexacyanoferrate (II) trihydraté 98,5-102,0, powder_yellow crystalline ACS
Potassium hexacyanoferrate (II) trihydraté
Potassium ferrocyanide trihydrate is a salt of the Fe(CN)₆4- ferrocyanide ion. Ferrocyanide is a reducing iron species, being oxidised from ferrous Fe(II) to ferric Fe(III).
It is used to determine the concentration of potassium permanganate, a compound often used in titrations based on redox reactions. Used in a mixture with potassium ferricyanide and phosphate buffered solution (PBS) to provide a buffer for X-Gal, which is used to cleave Beta-galactosidase, giving a bright blue visualisation where an antibody (or other molecule), conjugated to Beta-gal, has bonded to its target.
It is membrane impermeable and can be used to assess redox activity of membrane-bound structures and mechanisms of electron transport across membranes. This approach was employed to demonstrate that Potassium ferrocyanide performs reducing activation of membrane-bound dopamine β-monooxygenase from the exterior face of adrenal vesicles, providing elucidation of native reduction-activation mechanisms for dopamine β-monooxygenase regulation.
Formule: K₄Fe(CN)₆·3H₂O
Poids moléculaire: 422.39 g/mol
Point de fusion: 70 °C (lit.)                      
Densité: 7.87 g/cm³ (25 °C) (lit.)                      
Température de stockage: Température ambiante
Numéro MDL: MFCD00167023
Numéro CAS: 14459-95-1
Indice Merck: 12,07793
Commander maintenant
Résultats des tests de spécification
Assay 98,5 - 102,0%
Insoluble Matter ≤0,005%
Chloride ≤0,01%
Sulphate Passes Test