Imprimer
1,1'-Azobis(N,N-dimethylformamide), crystals_yellow
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): N1,N1,N2,N2-Tetramethyldiazene-1,2-dicarboxamide, Diamide
02101527.5 0210152701 0210152705 02101527.1
ICNA02101527.5EA 122 EUR
ICNA02101527.5 ICNA0210152701 ICNA0210152705 ICNA02101527.1
1,1'-Azobis(N,N-dimethylformamide), crystals_yellow
1,1'-Azobis(N,N-dimethylformamide)
Diamide is used as a thiol oxidising agent. It has also been used to titrate protein glutathiolation to discriminate from other oxidative protein modifications. Diamide treatment increased protein glutathiolation in a concentration-dependent manner and had comparably little effect on protein-protein disulphide formation.
Soluble in water.
Attention: Research Use Only (RUO).
Formule: C₆H₁₂N₄O₂
Poids moléculaire: 172.19 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00008318
Numéro CAS: 10465-78-8
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
Melting Point 104 - 115 °C
UV/Vis: λmax (water) 295 - 300 nm