Imprimer
Cleaning trolley Clino®, CR mini EM-GMP
Fournisseur: PFENNIG REINIGUNGSTECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3510170 3510160
PPSA3510170EA 0 EUR
PPSA3510170 PPSA3510160
Cleaning trolley Clino®, CR mini EM-GMP
Chariots
Impregnation system made of stainless steel suitable for cleanrooms and for high sensitive areas that require minimal space.
  • EasyMop GMP with system box certified for areas under sterille conditions
  • Highest efficiency and process reliability in single step technique
  • Basic module mini with high end finish and castors (100 mm)
The trolley is an high-end solution for the cleanroom cleaning in low classified and very small cleanrooms e.g. in pharmacies because it is suitable up to sterile areas. The EasyMop pre-preparation systems allows a validatable and reproducible cleaning and disinfection of surfaces. It can be stored under a workbench an easily stay in each room.
Certifications: IPA certificate for EasyMop GMP system box.
Informations de livraison: 1×system boxes EasyMop GMP and 1×stainless steel container (for EM-GMP1).