Imprimer
Water treatment units, Aqua purificator PG 8595
Fournisseur: MIELE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10087310 10087320
MIEL10087310EA 1620.78 EUR
MIEL10087310 MIEL10087320
Water treatment units, Aqua purificator PG 8595
Purificateurs d'eau Déioniseurs
Aqua purifier for two full or partial demineralisation cartridges. The casing made from white steel or stainless steel.
  • Compatible with PG 8583, PG 8593, PG 8583 series machines
Cabinet for two VE P 2000/VE P 2800 water demineralisation cartridges with conductivity meter, generally recommended quality for final rinse <19 µS/cm.