Imprimer
pH indicator, bromophenol blue tablets
Marques : Lovibond
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
511621 511620 221030
162623AEA 108 EUR
162623A LOVI511620 LOVI221030
pH indicator, bromophenol blue tablets
Kits de tests environnementaux
Indicator for pH range 2,8 to 4,4.
Can be used for measuring pH in conjunction with the Comparator 2000+ system and pH disc 2/1B