Imprimer
Disposable syringes
Fournisseur: THERMO NATIONAL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S7515-10 S7515-3 S7515-20 S7510-1
NSCAS7515-10EA 226 EUR
NSCAS7515-10 NSCAS7515-3 NSCAS7515-20 NSCAS7510-1
Disposable syringes
Seringues Seringues de chromatographie
These syringes are an ideal choice for all syringe filter applications. They feature a two-part construction using PP barrels and PE plungers. This eliminates the need for rubber or synthetic plunger gaskets and requires no silicone or oil lubricant in the barrel.
  • Non sterile
  • Bulk packed