Imprimer
Ring-type surface probes for Curvex dataloggers
Fournisseur: Industrial Physics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CX2066 CX2072 CX2065
SIMECX2066EA 201 EUR
SIMECX2066 SIMECX2072 SIMECX2065
Ring-type surface probes for Curvex dataloggers
Sondes de température
Ring-type surface probes to measure object temperature.
  • With aluminium ring at the tip for fast response
  • For use with Curvex oven data logger for powder coating ovens
  • Mandatory for Qualicoat, GSB or QIB accredited powder coaters to profile the curing process
Screw hole Ø: 5 mm