Imprimer
Screw caps for bottles with GL threads, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H987-05 H987-04 H987-03 H987-02 H987-01
BOHLH987-05EA 9.1 EUR
BOHLH987-05 BOHLH987-04 BOHLH987-03 BOHLH987-02 BOHLH987-01
Screw caps for bottles with GL threads, BOLA
Capuchons Bouchons pour flacons
Screw cap made of PBTP, red, with handy knurl and integrated PTFE/silicone gasket.
  • Very good chemical resistance
  • Temperature resistant from −50 to +140 °C
Suitable for bottles and vessels with GL threads.