Imprimer
Distillation apparatus set, general purpose
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2265400 2265800
HACH2265400EA 570 EUR
HACH2265400 HACH2265800
Distillation apparatus set, general purpose
Composants de distillation Pièces pour distillation
Fast set-up distillation apparatus for sample pretreatment. Distillation sets include only EPAapproved glassware.
  • Easy to set up, only required parts are delivered
  • Illustrated, step-by-step procedures simplify distillation
Adapts to a variety of test needs including all wastewater and water methods requiring sample pretreatment by distillation. Applications include fluoride, ammonia-nitrogen, phenols, selenium, volatile acids and albuminoid nitrogen.
Informations de commande: Includes distillation flask, J-tube, condenser, drip tube, erlenmeyer flask, and non-Hg thermometer. Arsenic and cyanide sets are available, these are used together with the general purpose distillation set.
Attention: Heater and support apparatus, sold separately, is required for distillation.