Imprimer
Signs, warning stands
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
241926 256702 256712 241930 092110 256281 241941
BRDY241926EA 55.7 EUR
BRDY241926 BRDY256702 BRDY256712 BRDY241930 BRDY092110 BRDY256281 BRDY241941
Signs, warning stands
Panneaux
These stable warning devices are easy to transport and suitable for the marking of hazardous zones.
  • Made of high-density polypropylene
  • Durable, yellow
  • No buckling or corrosion
  • Large note field is visible from over 9 meters
  • With default notices or unlabeled also available
Height: 660 mm