Imprimer
Lysing kits for Fastprep®-24 5G, Lysing Matrix A, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
116910050 116970050 116970025 116910500 116930050 116910100 116930005 116540423 116540427 116930025 116950010 116970100 116980010 116980001
ICNA116910050EA 225 EUR
ICNA116910050 ICNA116970050 ICNA116970025 ICNA116910500 ICNA116930050 ICNA116910100 ICNA116930005 ICNA116540423 ICNA116540427 ICNA116930025 ICNA116950010 ICNA116970100 ICNA116980010 ICNA116980001
Lysing kits for Fastprep®-24 5G, Lysing Matrix A, MP Biomedicals
Homogenizers Benchtop Homogenizers
Each impact-resistant 2 ml tube contains garnet matrix and one ¼ inch ceramic sphere. Extra ¼" ceramic spheres are packaged separately. Lysing Matrix A tubes have orange caps and are found in the FastDNA™ and FastDNA™ SPIN Kits. Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA, RNA, functional proteins and other metabolites and small molecules. This grinding matrix is chemically inert and will not bind nucleic acids.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable