Imprimer
Laboratory fume hood, PVC acid digestion, Protector®
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
141610021 141510021
LBCP141610021EA 0 EUR
LBCP141610021 LBCP141510021
Laboratory fume hood, PVC acid digestion, Protector®
Hottes et coffrets Hottes chimiques
The PVC acid digester Protector® laboratory hoods are strong acid resistant. Their liners are made up of unplasticised polyvinyl chloride.
  • Glacier white, dry powder-coated steel exterior with by-pass airflow design
  • Heat-welded unplasticised PVC liner with integral work surface and drainage trough
  • Built-in washdown system with internal piping and spray nozzles behind the baffles and remote control fixture on the left side
  • Removable front and side panels and front and interior access panels for access to plumbing and electrical wiring
  • Curved air foil allows air to sweep the work surface while Clean-Sweep™ openings pull inflow air from under the air foil