Imprimer
Containers, wide mouth, with screw caps, UV safe, Packo
Fournisseur: CURTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4303-57-101 4305-57-101 4310-57-101
CURT4303-57-101EA 5380 EUR
CURT4303-57-101 CURT4305-57-101 CURT4310-57-101
Containers, wide mouth, with screw caps, UV safe, Packo
Récipients de collecte d'échantillon Conteneurs à usage universel
Containers, made of black HDPE, UV safe, small volume with black screw lid and translucent innerseal.
  • Water vapour tight
  • Tamper evident
  • UN marking (1H2) for hazardous solids
  • Smooth inside
  • Food grade
For storage and sample shipment of UV sensitive powders powders, pastes and liquids. Easy to close and open with just a quarter-turn.