Imprimer
Temperature stabilisation jacket
Fournisseur: SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
285405532 285405508 285405549 285405516
SCOI285405532EA 22.4 EUR
SCOI285405532 SCOI285405508 SCOI285405549 SCOI285405516
Temperature stabilisation jacket
Contrôleurs Contrôleurs de température
The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
  • Support plate to facilitate the use
  • Quick-action seal to simplify the changing viscometers
  • Additional hole in the support plate so that a control thermometer can fit
Suitable for use with Ubbelohde viscometers.