Imprimer
Density hydrometers, M100
Fournisseur: Amarell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H809616 H809610 H809612 H809604 H809608 H809622 H809602 H809618 H809620 H809604/WC H809608/WC H809610/WC H809612/WC H809614/WC H809616/WC H809618/WC H809620/WC H809602/WC H809614 H809624 H809626 H809604/DKD3 H809606 H809600 H809610/DKD3 H809612/DKD3 H809618DKD3 H809616DKD3
614-0534EA 44.8 EUR
614-0534 AMARH809610 AMARH809612 AMARH809604 AMARH809608 AMARH809622 AMARH809602 AMARH809618 AMARH809620 AMARH809604/WC AMARH809608/WC AMARH809610/WC AMARH809612/WC AMARH809614/WC AMARH809616/WC AMARH809618/WC AMARH809620/WC AMARH809602/WC AMARH809614 AMARH809624 AMARH809626 AMARH809604/DKD3 AMARH809606 AMARH809600 AMARH809610/DKD3 AMARH809612/DKD3 AMARH809618DKD3 AMARH809616DKD3
Density hydrometers, M100
Hydromètres
Hydrometers designed with paper scale, colour code, type designation and weighted body.
  • Round capillary
  • Accuracy of ±1 °C
  • Thermometer in the hydrometer
Suitable for government verification with (or without) mercury thermometer.
Certifications: DIN 12791.
Cdt: In a transparent plastic case.