Imprimer
Cleaning brushes for butyrometer
Fournisseur: GERBER FUNKE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3324 3325
614-2036EA 8.2 EUR
614-2036 614-2037
Cleaning brushes for butyrometer
Brosses
Cleaning brush for butyrometer body
  • Total length: 270 mm
  • Brush length: 100 mm