Imprimer
Anaerobic vessels with Petri dish insert rack
Fournisseur: schuett-biotec GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3380102 3380202 3880600 3385102 3385302 3880300 3380902
SCUT3380102EA 919 EUR
SCUT3380102 SCUT3380202 SCUT3880600 SCUT3385102 SCUT3385302 SCUT3880300 SCUT3380902
Anaerobic vessels with Petri dish insert rack
Anaerobic Systems
For the cultivation of anaerobic or microaerophilic microorganisms in a defined, rapidly developing and optimal gas atmosphere. Racks (optional): For Petri dishes, microtiter plates or test tubes. Equipped with holders for anaerobic systems.
  • The container is unbreakable and non-aging and made of rustproof, robust stainless steel or transparent polycarbonate
  • The vessel lid is made of UV-stable plastic or transparent polycarbonate