Imprimer
Cell factories, Nunc™ high density
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
169102 169118 169160
NUNC169102EA 1162 EUR
NUNC169102 NUNC169118 NUNC169160
Cell factories, Nunc™ high density
Bioréacteurs
Thermo Scientific™ Nunc™ High Density Cell Factory™ system offers greater surface area and yield than the standard Cell Factory™ system, or similar multi-tray systems for adherent cell culture. It offers an efficient, economic option for increasing production capacity and cell culture yield in the same footprint.
  • Certified Nunclon™∆ surface treatment ensures excellent conditions for cell attachment and growth
  • Increased yield resulting from 30% more surface area within the standard Cell Factory™ footprint
  • Increased labour and handling efficiencies by achieving more output in a single process run
  • Increase manufacturing capacity without capital investment
  • Lessen material usage and reduce decontamination and waste disposal costs
Its has additional layers. Cell factory systems are designed to optimise any process by increasing output, while maintaining the same manufacturing footprint as with the standard cell factory system.
Consume less and reduce decontamination and waste disposal costs.