Imprimer
Wall cleaning tool
Fournisseur: PFENNIG REINIGUNGSTECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3580038 3580033 3580039
PPSA3580038EA 0 EUR
PPSA3580038 PPSA3580033 PPSA3580039
Wall cleaning tool
Serpillières
Wall cleaning tool for wall and ceiling cleaning in hygienically sensitve areas.
  • Frame completely made of stainless steel
  • Mops are 100% polyester microfiber filament yarn
  • Autoclavable, disinfectant proof
  • Mops have high-quality stainless steel push buttons for fitting with frame
This mop frame with locking screw is suitable with mop covers Micro Sicuro CR/A and Mop CR for wall cleaning tool.
Cdt: Mops are prewashed and single double packaged.