Imprimer
Joint scrubber/brush for cleanrooms
Fournisseur: PFENNIG REINIGUNGSTECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3590600 3590510
PPSA3590600EA 39.3 EUR
PPSA3590600 PPSA3590510
Joint scrubber/brush for cleanrooms
Brosses
Joint scrubber/brush for gaps in hygienic sensitive areas.
  • Material: PP (body), PBT (stock)
  • Unslit or spouted fibre ends
  • Medium-hard bristle material
  • Heat-resistant up to 140 °C
Colour: White