Imprimer
Sharps Containers
Fournisseur: HEATHROW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HS120178 HS120177
113-8044EA 130 EUR
113-8044 HEATHS120177
Sharps Containers
Poubelles Collecteurs de déchets tranchants
This sharps container provides safe disposal of syringes, needles and other small sharps. Easy to use and transport.
  • Stackable to reduce storage space before use
  • With a clear lid to view fill level
  • Slide access lid for temporary and final closures for safety and security
  • Built-in needle removal port, preventing direct contact with sharps
  • Compatible with the Droplet® Blood Collection Tray