Imprimer
Immersion circulators, AC150
Fournisseur: THERMO ELECTRON LED
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1550021 1550001B
HAKA1550021EA 3450 EUR
HAKA1550021 HAKA1550001B
Immersion circulators, AC150
Thermostats à circulation Systèmes à circulation ouverte Contrôleurs à thermostat
AC150 Immersion circulators are ideal for applications that require greater pumping performance with features such as ramp programming, application alarms and temperature ranges from −50 to 150 °C.
  • Automatic controller shutdown at detection of excessive high temperature, low liquid level or motor overload
  • Programmable application temperature alarm with user selected alarm, go-safe-state or shut off option
  • Five programmable set point temperatures and self-tuning PID controller for optimized temperature control
  • Low liquid level alarm for Safety Class III and RTA (Real Temperature Adjustment) for calibration
  • Powerful force and suction pump for external open and closed applications
Also features USB communication port and options for RS232, RS485, Ethernet/LAN, Analog I/O and internal or external temperature control mode (Remote Sensor, NAMUR type). Acoustic and optical alarm with auto restart feature after power failure.
Informations de livraison: It includes hose barbs with bridge (8 and 12 mm), pump plug for external circulation and power cord (1,8 m).