Imprimer
Mesotherapy needles
Fournisseur: EUROMEDIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
102100 102101
EUMD102100EA 89.3 EUR
EUMD102100 EUMD102101
Mesotherapy needles
Aiguilles pour seringues
Mesotherapy needles are finer and smaller, adapted to treatment of venous circulatory problems and water retention.
  • By the conformation of their bevel (triple chip technology), stainless steel needles allow painless sessions
  • Sterile, single use and latex free
  • Fit over a Luer cone or Luer lock
  • With translucent base (Ø 0,3 mm)