Imprimer
Turbidity standards, high range 3000 to 4000 NTU
Marques : REAGECON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CRS-4000-100 CRS-4000-500 CRSR-4000-500 CRSR-4000-100
REGECRS-4000-100EA 518 EUR
REGECRS-4000-100 REGECRS-4000-500 REGECRSR-4000-500 REGECRSR-4000-100
Turbidity standards, high range 3000 to 4000 NTU
Étalons Étalons de turbidité
Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
  • Non toxic and non carcinogenic microspheres
  • Highly accurate with well defined spherical shape
  • Traceable to NIST
  • US EPA approved
  • Ready to use- particles stay in suspension