Imprimer
Turbidity standards, low range 2 to 10 NTU
Marques : REAGECON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CRS-10-500 CRS-2-100 CRSR-2-500 CRS-5-100 CRS-5-500 CRSR-10-500 CRSR-2-100 CRSR-4-100 CRSR-4-500 CRSR-5-100 CRSR-5-500 CRSR-10-100 CRS-2-500
REGECRS-10-500EA 634 EUR
REGECRS-10-500 REGECRS-2-100 REGECRSR-2-500 REGECRS-5-100 REGECRS-5-500 REGECRSR-10-500 REGECRSR-2-100 REGECRSR-4-100 REGECRSR-4-500 REGECRSR-5-100 REGECRSR-5-500 REGECRSR-10-100 REGECRS-2-500
Turbidity standards, low range 2 to 10 NTU
Étalons Étalons de turbidité
Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
  • Non toxic and non carcinogenic microspheres
  • Highly accurate with well defined spherical shape
  • Traceable to NIST
  • US EPA approved
  • Ready to use- particles stay in suspension