Imprimer
Industrial gloves, uvex phynomic wet
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6006009 6006008 6006007 6006010
UVEX6006009EA 64.8 EUR
UVEX6006009 UVEX6006008 UVEX6006007 UVEX6006010
Industrial gloves, uvex phynomic wet
Gants
This polyamide/elastane glove with Aqua polymer foam coating on palm and fingertips is particularly suitable for damp/oily working conditions.
  • Excellent dexterity
  • Good dry and wet grip
  • High level of breathability ensures optimum wearer climate and comfort
  • Water-repellent coating and liner
  • Oil/emulsions roll off, oil is absorbed
  • Skin safe, dermatologically approved
Ideal for use in more demanding indoor and outdoor areas of application. Typical applications include: Fine assembly work/assembly work, general maintenance work and precision work.
Certifications: EN 388:2016 (3131X)
OEKO-TEX® Standard 100