Imprimer
Biopsy punches
Fournisseur: LABONORD MISCELLANEOUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
48201 48301 48351 48401 48501 48801 48601
LBMI48201EA 130 EUR
LBMI48201 LBMI48301 LBMI48351 LBMI48401 LBMI48501 LBMI48801 LBMI48601
Biopsy punches
Poinçons de biopsie
This biopsy punch makes it possible to perform skin biopsies in various locations with great precision and minimal tissue trauma. The stainless steel blade is ultra sharp thanks to high-precision micro-grinding technology.
  • Ribbed handle ensures a safe grip and optimal control during specimen collection
  • Size information on each punch
  • Sterile packed disposable punch makes grinding and sterilising unnecessary