Imprimer
Acidic cleaner and neutraliser, deconex® 26 MINERALACID
Fournisseur: Borer Chemie
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
502200.00-KK5R 502200.00-K10R 502200.00-F10R
148-5261EA 71.1 EUR
148-5261 148-5262 DECO502200.00-F10R
Acidic cleaner and neutraliser, deconex® 26 MINERALACID
Produits nettoyants Détergents et Nettoyants de Surfaces
Liquid, acidic cleaning concentrate and neutraliser for automated cleaning of laboratory glassware and reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • Neutralises carried over alkaline residues and substantially reduces rinsing time and consumption of water
  • In combination with all alkaline deconex® cleaning products, can be also used as a precleaner to remove acid soluble deposits
  • Removes lime residues in laboratory washers
  • Suitable for use in laboratories in the dairy industry
  • EDTA-, NTA-, chlorine- and surfactant-free, no release of surfactants into waste water