Imprimer
Cryopreservation cane
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4000211 4000217
BARN4000211EA 21.9 EUR
BARN4000211 BARN4000217
Cryopreservation cane
Dewars et récipients cryogéniques
These cane systems made of aluminium are used to store cell, tissues or other biological samples.
  • Cost effective
  • Durable aluminum construction and vacuum insulation
  • Narrow-mouth design minimises LN2 evaporation
  • Secure locking clasp
Colour: Silver