Imprimer
Business socks
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
900016/35-38 900016/39-42 900016/43-46 900016/47-50
ELTN900016/35-38EA 6.3 EUR
ELTN900016/35-38 ELTN900016/39-42 ELTN900016/43-46 ELTN900016/47-50
Business socks
Tenue de travail
Elegant and comfortable business socks made of 98% nylon and 2% elastane.
  • Highly elastic, pressure-free cuffs
  • Heel, toe and sole reinforcements
  • Anatomically shaped foot bed is designed to fit the right and left foot
  • Washed antistatically