Imprimer
Microscope slides for printers
Fournisseur: KLINIPATH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
631-1231EA 158 EUR
631-1231 631-1232 631-1233 631-1234 631-1235
Microscope slides for printers
Lames Lames de microscopes
Microscope slides for slide printers allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
  • Frosted ends
  • 90° grounded edges
  • Finished clipped corners of 1mm